Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAGKRSGWSR AALLQLLLGV NLVVMPPTQA RSLRFVTLLY RHGDRSPVKT YPKDPYQEEE WPQGFGQLTK EGMLQHWELG QALRQRYHGF LNTSYHRQEV YVRSTDFDRT LMSAEANLAG LFPPNGMQRF NPNISWQPIP VHTVPITEDR LLKFPLGPCP RYEQLQNETR QTPEYQNESS RNAQFLDMVA NETGLTDLTL ETVWNVYDTL FCEQTHGLRL PPWASPQTMQ RLSRLKDFSF RFLFGIYQQA EKARLQGGVL LAQIRKNLTL MATTSQLPKL LVYSAHDTTL VALQMALDVY NGEQAPYASC HIFELYQEDS GNFSVEMYFR NESDKAPWPL SLPGCPHRCP LQDFLRLTEP VVPKDWQQEC QLASGPADTE VIVALAVCGS ILFLLIVLLL TVLFRMQAQP PGYRHVADGE DHA
This gene encodes the beta subunit of lysosomal acid phosphatase (LAP). LAP is chemically and genetically distinct from red cell acid phosphatase. The encoded protein belongs to a family of distinct isoenzymes which hydrolyze orthophosphoric monoesters to alcohol and phosphate. Mutations in this gene or in the related alpha subunit gene cause acid phosphatase deficiency. Multiple alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LAP; Lysosomal acid phosphatase
Gene Aliases: ACP2; LAP
UniProt ID: (Human) P11117
Entrez Gene ID: (Human) 53
Molecular Function:
phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support