Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MITGVFSMRL WTPVGVLTSL AYCLHQRRVA LAELQEADGQ CPVDRSLLKL KMVQVVFRHG ARSPLKPLPL EEQVEWNPQL LEVPPQTQFD YTVTNLAGGP KPYSPYDSQY HETTLKGGMF AGQLTKVGMQ QMFALGERLR KNYVEDIPFL SPTFNPQEVF IRSTNIFRNL ESTRCLLAGL FQCQKEGPII IHTDEADSEV LYPNYQSCWS LRQRTRGRRQ TASLQPGISE DLKKVKDRMG IDSSDKVDFF ILLDNVAAEQ AHNLPSCPML KRFARMIEQR AVDTSLYILP KEDRESLQMA VGPFLHILES NLLKAMDSAT APDKIRKLYL YAAHDVTFIP LLMTLGIFDH KWPPFAVDLT MELYQHLESK EWFVQLYYHG KEQVPRGCPD GLCPLDMFLN AMSVYTLSPE KYHALCSQTQ VMEVGNEE
Lysophosphatidic acid phosphatase type 6 (ACP6), also designated acid phosphatase-like protein 1 (ACPL1) or lysophosphatidic acid phosphatase (LPAP), is a 428 amino acid secreted protein that hydrolyzes lysophosphatidic acid to monoacylglycerol. ACP6 is highly expressed in kidney, heart, small intestine, muscle, liver, prostate, testis, ovary and exists as two isoforms as a result of alternative splicing events. The gene encoding ACP6 maps to human chromosome 1, the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene of human chromosome 1, which encodes lamin A. Stickler syndrome, Parkinsons, Gaucher disease, familial adenomatous polyposis and Usher syndrome are also associated with chromosome 1. Aberrations in chromosome 1 are found in a variety of cancers including head and neck cancer, malignant melanoma and multiple myeloma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3.1.3.2; Acid phosphatase 6, lysophosphatidic; acid phosphatase like 1; Acid phosphatase-like protein 1; Lysophosphatidic acid phosphatase type 6; PACPL1
Gene Aliases: 5730559A09Rik; ACP6; ACPL1; AU022842; LPAP; mPACPL1; PACPL1; UNQ205/PRO231
UniProt ID: (Human) Q9NPH0, (Mouse) Q8BP40
Entrez Gene ID: (Human) 51205, (Mouse) 66659
Molecular Function:
phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support