Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VAEDKLLRF PMRSCPRYHE LLREATEAAE YQEALEGWTG FLSRLENFTG LSLVGEPLRR AWKVLDTLMC QQAHGLPLPA WASPDVLRTL AQISALDIGA HVGPPRAAEK AQLTGGILLN AILANFSRVQ RLGLPLKMVM YSAHDSTLLA LQGALGLYDG HTPPYAACLG FEFRKHLGNP AKDGGNVTVS LFYRNDSAHL PLPLSLPGCP APCPLGRFYQ LTA (146-367 aa encoded by BC146506)
Acid phosphatases are enzymes capable of hydrolyzing orthophosphoric acid esters in an acid medium. This gene is up-regulated by androgens and is down-regulated by estrogens in the prostate cancer cell line. This gene exhibits a lower level of expression in testicular cancer tissues than in normal tissues. The protein encoded by this gene has structural similarity to prostatic and lysosomal acid phosphatases. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acid phosphatase 4; Testicular acid phosphatase
Gene Aliases: ACP4; ACPT; EG546967; Gm1432
UniProt ID: (Human) Q9BZG2, (Mouse) D3YTS9
Entrez Gene ID: (Human) 93650, (Mouse) 100503991
Molecular Function:
phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support