Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AMSVTLEQN LTCPGSDLKP FTTRLADYLV LAKVRQALGF AKCQKNFYGA APMMAETQHF FLGLNIRLYA GYGLSETSGP HFMSSPYNYR LYSSGKLVPG CRVKLVNQDA EGIGEICLWG RTIFMGYLNM EDKTCEAIDE EGWLHTGDAG RLDADGFLYI TGRLKELIIT AGGENVPPVP IEEAVKMELP IISNAMLIGD QRKFLSMLLT LKCTLDPDTS DQTDNLTEQA VEFCQRVGSR ATTVSEIIEK KDEAVYQAIE EGIRRVNMNA AARPYHIQKW AILE (404-686 aa encoded by B C009289)
The protein encoded by this gene possesses long-chain acyl-CoA synthetase activity. It is thought to play a central role in brain very long-chain fatty acids metabolism and myelinogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acyl-CoA synthetase bubblegum family member 1; bubblegum; Gonadotropin-regulated long chain acyl CoA synthetase; gonadotropin-regulated long chain acyl-CoA synthetase; GR-LACS; hBG1; Lipidosin; lipidosis-related protein lipidosin; lipodisin; Long-chain-fatty-acid--CoA ligase ACSBG1; mBG1; very long-chain acyl-CoA synthetase
Gene Aliases: ACSBG1; BG; BG1; BGM; E230019G03Rik; GR-LACS; Grlacs; KIAA0631; LPD; mKIAA0631; R75185
UniProt ID: (Human) Q96GR2, (Mouse) Q99PU5, (Rat) Q924N5
Entrez Gene ID: (Human) 23205, (Mouse) 94180, (Rat) 171410
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support