Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPRNSGAGYG CPHGDPSMLD SRETPQESRQ DMIVRTTQEK LKTSSLTDRQ PLSKESLNHA LELSVPEKVN NAQWDAPEEA LWTTRADGRV RLRIDPSCPQ LPYTVHRMFY EALDKYGDLI ALGFKRQDKW EHISYSQYYL LARRAAKGFL KLGLKQAHSV AILGFNSPEW FFSAVGTVFA GGIVTGIYTT SSPEACQYIA YDCCANVIMV DTQKQLEKIL KIWKQLPHLK AVVIYKEPPP NKMANVYTME EFMELGNEVP EEALDAIIDT QQPNQCCVLV YTSGTTGNPK GVMLSQDNIT WTARYGSQAG DIRPAEVQQE VVVSYLPLSH IAAQIYDLWT GIQWGAQVCF AEPDALKGSL VNTLREVEPT SHMGVPRVWE KIMERIQEVA AQSGFIRRKM LLWAMSVTLE QNLTCPGSDL KPFTTRLADY LVLAKVRQAL GFAKCQKNFY GAAPMMAETQ HFFLGLNIRL YAGYGLSETS GPHFMSSPYN YRLYSSGKLV PGCRVKLVNQ DAEGIGEICL WGRTIFMGYL NMEDKTCEAI DEEGWLHTGD AGRLDADGFL YITGRLKELI ITAGGENVPP VPIEEAVKME LPIISNAMLI GDQRKFLSML LTLKCTLDPD TSDQTDNLTE QAMEFCQRVG SRATTVSEII EKKDEAVYQA IEEGIRRVNM NAAARPYHIQ KWAILERDFS ISGGELGPTM KLKRLTVLEK YKGIIDSFYQ EQKM
The protein encoded by this gene possesses long-chain acyl-CoA synthetase activity. It is thought to play a central role in brain very long-chain fatty acids metabolism and myelinogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acyl-CoA synthetase bubblegum family member 1; bubblegum; hBG1; Lipidosin; Long-chain-fatty-acid--CoA ligase ACSBG1; very long-chain acyl-CoA synthetase
Gene Aliases: ACSBG1; BG; BG1; BGM; GR-LACS; KIAA0631; LPD
UniProt ID: (Human) Q96GR2
Entrez Gene ID: (Human) 23205
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support