Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FNGKKALEA ISRERGTFLY GTPTMFVDIL NQPDFSSYDI STMCGGVIAG SPAPPELIRA IINKINMKDL VVAYGTTENS PVTFAHFPED TVEQKAESVG RIMPHTEARI MNMEAGTLAK LNTPGELCIR GYCVMLGYWG EPQKTEEAVD QDKWYWTGDV ATMNEQGFCK IVGRSKDMII RGGENIYPAE LEDFFHTHPK VQEVQVVGVK DDRMGEEICA CIRLKDGEET TVEEIKAFCK GKISHFKIPK YIVFVTNYPL TISGKIQKFK LREQMERHLN L (336-615 aa encoded by BC012053)
Acyl-CoA synthases catalyze the initial reaction in fatty acid metabolism, by forming a thioester with CoA (PubMed:17762044). Has some preference toward medium-chain substrates (PubMed:17762044). Plays a role in adipocyte differentiation (PubMed:16380219). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acyl-CoA synthetase family member 2, mitochondrial; Medium-chain acyl-CoA ligase ACSF2, mitochondrial; PPARG binding, long chain fatty acid acyl Co-A ligase like
Gene Aliases: ACSF2; ACSMW; AVYV493; UNQ493/PRO1009
UniProt ID: (Human) Q96CM8, (Mouse) Q8VCW8, (Rat) Q499N5
Entrez Gene ID: (Human) 80221, (Mouse) 264895, (Rat) 619561
Molecular Function:
ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support