Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MRPWLRHLV LQALRNSRAF CGSHGKPAPL PVPQKIVATW EAISLGRQLV PEYFNFAHDV LDVWSRLEEA GHRPPNPAFW WVNGTGAEIK WSFEELGKQS RKAANVLGGA CGLQPGDRMM LVLPRLPEWW LVSVACMRTG TVVIPGVTQL TEKDLKYRLQ ASRAKSIITS DSLAPRVDAI SAECPSLQTK LLVSDSSRPG WLNFRELLR (1-208 aa encoded by BC016703 )
Catalyzes the activation of fatty acids by CoA to produce an acyl-CoA, the first step in fatty acid metabolism. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acyl-CoA synthetase medium-chain family member 5; Acyl-coenzyme A synthetase ACSM5, mitochondrial
Gene Aliases: Aa2-174; ACSM5; C730019D22; C730027J19Rik; Cc1-38; MACS3; RGD1309578
UniProt ID: (Human) Q6NUN0, (Rat) Q6AYT9, (Mouse) Q8BGA8
Entrez Gene ID: (Human) 54988, (Rat) 361637, (Mouse) 272428
Molecular Function:
ligase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support