Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MCEEETTALV CDNGSGLCKA GFAGDDAPRA VFPSIVGRPR HQGVMVGMGQ KDSYVGDEAQ SKRGILTLKY PIEHGIITNW DDMEKIWHHS FYNELRVAPE EHPTLLTEAP LNPKANREKM TQIMFETFNV PAMYVAIQAV LSLYASGRTT GIVLDSGDGV THNVPIYEGY ALPHAIMRLD LAGRDLTDYL MKILTERGYS FVTTAEREIV RDIKEKLCYV ALDFENEMAT AASSSSLEKS YELPDGQVIT IGNERFRCPE TLFQPSFIGM ESAGIHETTY NSIMKCDIDI RKDLYANNVL SGGTTMYPGI ADRMQKEITA LAPSTMKIKI IAPPERKYSV WIGGSILASL STFQQMWISK PEYDEAGPSI VHRKCF
Actin exists as a ubiquitous protein involved with filament formation that make up large portions of the cytoskeleton. Actin filaments interact with myosin to assist in muscle contraction as well as aiding in cell motility and cytokinesis. In vertebrates there are three groups of actin isoforms: alpha, beta and gamma. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. The beta and gamma actins co-exists in most cell types as components of the cytoskeleton and as mediators of internal cell motility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Actin, gamma-enteric smooth muscle; actin-like protein; Alpha-actin-3; Gamma-2-actin; Smooth muscle gamma-actin
Gene Aliases: ACT; ACTA3; ACTE; ACTG2; ACTL3; ACTSG; VSCM
UniProt ID: (Human) P63267
Entrez Gene ID: (Human) 72
Molecular Function:
actin and actin related protein
actin or actin-binding cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support