Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MASRTVIIDH GSGFLKAGTA GWNEPQMVFP NIVNYLPCKE NPGPSYARRR VSLGIDICHP DTFSYPIERG RILNWEGVQY LWSFVLENHR REQEVPPVII TETPLREPAD RKKMLEILFE LLHVPSVLLA DQLQMSLYAS GLLTGVVVDS GYGLTRVQPF HQGRPLPASG KTLEFAGQDL SAYLLKSLFK EDCDRRCLFQ LETVAVTQMN KCYVPQNLGE ALDFRERQQS ALDESNTYQL PDGSRVELTP MQRVAPEMFF SPQVFEQPGP SIPRAIVESV ESCEISLRPL LVSHVMACGG NTLYPGFTKR LFRELMGDHV SSTKATVWEG SNRNFSVWLG ASVVAHLSTY QSEWMSREEY GEHMRM
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin like protein; actin-like 8; Actin-like protein 8; Cancer/testis antigen 57; CT57
Gene Aliases: ACTL8; CT57
UniProt ID: (Human) Q9H568
Entrez Gene ID: (Human) 81569
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support