Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAGSLPPCVV DCGTGYTKLG YAGNTEPQFI IPSCIAIRES AKVVDQAQRR VLRGVDDLDF FIGDEAIDKP TYATKWPIRH GIIEDWDLME RFMEQVVFKY LRAEPEDHYF LMTEPPLNTP ENREYLAEIM FESFNVPGLY IAVQAVLALA ASWTSRQVGE RTLTGIVIDS GDGVTHVIPV AEGYVIGSCI KHIPIAGRDI TYFIQQLLRE REVGIPPEQS LETAKAIKEK YCYICPDIVK EFAKYDVDPR KWIKQYTGIN AINQKKFVID VGYERFLGPE IFFHPEFANP DFMESISDVV DEVIQNCPID VRRPLYKNVV LSGGSTMFRD FGRRLQRDLK RVVDARLRLS EELSGGRIKP KPVEVQVVTH HMQRYAVWFG GSMLASTPEF FQVCHTKKDY EEYGPSICRH NPVFGVMS
ACTR3B plays a role in the organization of the actin cytoskeleton.ACTR3B may function as ATP-binding component of the Arp2/3 complex which is involved in regulation of actin polymerization and together with an activating nucleation-promoting factor (NPF) mediates the formation of branched actin networks. ACTR3B may decrease the metastatic potential of tumors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Actin-like protein 3B; actin-related protein 3-beta; Actin-related protein 3B; actin-related protein Arp11; Actin-related protein ARP4; ARP3 actin-related protein 3 homolog B; ARP3-beta; DKFZp686O24114
Gene Aliases: ACTR3B; ARP11; ARP3BETA; ARP4
UniProt ID: (Human) Q9P1U1
Entrez Gene ID: (Human) 57180
Molecular Function:
actin and actin related protein
actin or actin-binding cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support