Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: CIKHIPIAGR DITYFIQQLL REREVGIPPE QSLETAKAIK EKYCYICPDI VKEFAKYDVD PRKWIKQYTG INAINQKKFV IDVGYERFLG PEIFFHPEFA NPDFMESISD VVDEVIQNCP IDVRRPLYKN VVLSGGSTMF RDFGRRLQRD LKRVVDARLR LSEELSGGRI KPKPVEVQVV THHMQRYAVW FGGSMLASTP EFFQVCHTKK DYEEYGPSIC RHNPVFGVMS; Positive Samples: HeLa, NCI-H125, Mouse brain; Cellular Location: Cell projection, Cytoplasm, cytoskeleton
ACTR3B plays a role in the organization of the actin cytoskeleton.ACTR3B may function as ATP-binding component of the Arp2/3 complex which is involved in regulation of actin polymerization and together with an activating nucleation-promoting factor (NPF) mediates the formation of branched actin networks. ACTR3B may decrease the metastatic potential of tumors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Actin-like protein 3B; actin-related protein 3-beta; Actin-related protein 3B; actin-related protein Arp11; Actin-related protein ARP4; ARP3 actin-related protein 3 homolog B; ARP3-beta; DKFZp686O24114
Gene Aliases: 9630005C02; ACTR3B; ARP11; Arp3b; ARP3BETA; ARP4; AW047569
UniProt ID: (Human) Q9P1U1, (Mouse) Q641P0
Entrez Gene ID: (Human) 57180, (Mouse) 242894
Molecular Function: actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support