Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGAAAKLAFA VFLISCSSGA ILGRSETQEC LFFNANWEKD RTNQTGVEPC YGDKDKRRHC FATWKNISGS IEIVKQGCWL DDINCYDRTD CVEKKDSPEV YFCCCEGNMC NEKFSYFPEM EVTQPTSNPV TPKPPYYNIL LYSLVPLMLI AGIVICAFWV YRHHKMAYPP VLVPTQDPGP PPPSPLLGLK PLQLLEVKAR GRFGCVWKAQ LLNEYVAVKI FPIQDKQSWQ NEYEVYSLPG MKHENILQFI GAEKRGTSVD VDLWLITAFH EKGSLSDFLK ANVVSWNELC HIAETMARGL AYLHEDIPGL KDGHKPAISH RDIKSKNVLL KNNLTACIAD FGLALKFEAG KSAGDTHGQV GTRRYMAPEV LEGAINFQRD AFLRIDMYAM GLVLWELASR CTAADGPVDE YMLPFEEEIG QHPSLEDMQE VVVHKKKRPV LRDYWQKHAG MAMLCETIEE CWDHDAEARL SAGCVGERIT QMQRLTNIIT TEDIVTVVTM VTNVDFPPKE SSL
Activin A receptor kinase (ACVR2A) is a type II member of the TGF-beta family of receptor Serine/Threonine kinases. ACVR2A is involved in the activin and myostatin signaling pathway. (ACVR2A Aliases: ACVR2, ACTRII). These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling, and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. Type II receptors are considered to be constitutively active kinases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activin A receptor type IIA; activin A receptor, type IIA; Activin receptor type IIA; Activin receptor type-2A; ACTR-IIA
Gene Aliases: ACTRII; ACVR2; ACVR2A
UniProt ID: (Human) P27037
Entrez Gene ID: (Human) 92
Molecular Function:
serine/threonine protein kinase receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support