Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MCSLPVPRE PLRRVAVTGG THGNEMSGVY LARHWLHAPA ELQRASFSAV PVLANPAATS GCRRYVDHDL NRTFTSSFLN SRPTPDDPYE VTRARELNQL LGPKASGQAF DFVLDLHNTT ANMGTCLIAK SSHEVFAMHL CRHLQLQYPE LSCQVFLYQR SGEESYNLDS VAKNGLGLEL GPQPQGVLRA DIFSRMRTLV ATVLDFIELF NQGTAFPAFE MEAYRPVGVV DFPRTEAGHL AGTVHPQLQD RDFQPLQPGA PIFQMFSGED LLYEGESTVY PVFINEAAYY EKGVAFVQTE KFTFTVPAMP ALTPAPSPAS (1-319 aa encoded by BC008689)
ACY3 belongs to the aspA/astE family. Aspartoacylase subfamily. Catalytic activity: N-acyl-L-aspartate + H(2)O = a carboxylate + L-aspartate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AAIII; ACY-3; Acylase III; Aminoacylase III; Aminoacylase-3; aspartoacylase (aminoacylase) 3; aspartoacylase (aminocyclase) 3; aspartoacylase 3; Aspartoacylase-2; aspartoacylase-3; HCBP1; HCV core-binding protein 1; Hepatitis C virus core-binding protein 1; N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming)
Gene Aliases: 0610006H10Rik; AA3; AAIII; ACY-3; ACY3; ASPA2; AW107362; HCBP1; RGD1305394
UniProt ID: (Human) Q96HD9, (Rat) Q5M876, (Mouse) Q91XE4
Entrez Gene ID: (Human) 91703, (Rat) 293653, (Mouse) 71670
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support