Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: GHPTVHSETK YVELIVINDH QLFEQMRQSV VLTSNFAKSV VNLADVIYKE QLNTRIVLVA METWADGDKI QVQDDLLETL ARLMVYRREG LPEPSDATHL FSGR
Probable ligand for integrin in the brain. This is a non catalytic metalloprotease-like protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a disintegrin and metalloproteinase; ADAM; ADAM 11; ADAMs; Disintegrin and metalloproteinase domain-containing protein 11; MDC; metalloendopeptidases; Metalloproteinase-like, disintegrin-like, and cysteine-rich protein
Gene Aliases: ADAM11; MDC
UniProt ID: (Human) O75078
Entrez Gene ID: (Human) 4185
Molecular Function: metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support