Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
ADAM12 is a member of the ADAM (a disintegrin and metalloprotease-like domain) family. Two forms of ADAM12 have been described: ADAM12S and ADAM12L (short and long forms). The short form is a soluble form lacking the transmembrane and cytoplasmic domains. The short form of ADAM12 was reported to provoke myogenesis, and the lack of cytoplasmic domain suggests different regulation pathways (although both forms can be expressed in the same tissue).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a disintegrin and metalloproteinase; ADAM; ADAM 12; ADAMs; Disintegrin and metalloproteinase domain-containing protein 12; Meltrin-alpha; metalloendopeptidases; metalloprotease-disintegrin 12 transmembrane; PRO545; RP11-295J3.5; UNQ346
Gene Aliases: ADAM12; ADAM12-OT1; CAR10; MCMP; MCMPMltna; MLTN; MLTNA; UNQ346/PRO545
UniProt ID: (Human) O43184
Entrez Gene ID: (Human) 8038
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support