Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Samples: Mouse heart, Mouse kidney, Mouse brain; Cellular Location: Cell junction, Cell projection, Cytoplasmic vesicle, Endomembrane system, Single-pass type I membrane protein, acrosome, adherens junction, cilium, flagellum, secretory vesicle
Immunogen sequence: QPAAPLCLQT ANTRGNAFGS CGRNPSGSYV SCTPRDAICG QLQCQTGRTQ PLLGSIRDLL WETIDVNGTE LNCSWVHLDL GSDVAQPLLT LPGTACGPGL VCIDHRCQRV DLLGAQECRS KCHGHGVCDS NRHCYCEEGW APPDCTTQLK ATSSLTTGLL L
The protein encoded by this gene is a member of the ADAM (a disintegrin and metalloproteinase) protein family. ADAM family members are type I transmembrane glycoproteins known to be involved in cell adhesion and proteolytic ectodomain processing of cytokines and adhesion molecules. This protein contains multiple functional domains including a zinc-binding metalloprotease domain, a disintegrin-like domain, as well as a EGF-like domain. Through its disintegrin-like domain, this protein specifically interacts with the integrin beta chain, beta 3. It also interacts with Src family protein-tyrosine kinases in a phosphorylation-dependent manner, suggesting that this protein may function in cell-cell adhesion as well as in cellular signaling. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a disintegrin and metallopeptidase domain 15 (metargidin); a disintegrin and metalloprotease domain 15 (metargidin); a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 15 (metargidin); AD56; ADAM; ADAM 15; ADAMs; CRII-7; disintegrin and metalloprotease 15; Disintegrin and metalloproteinase domain-containing protein 15; MDC-15; metalloendopeptidases; Metalloprotease RGD disintegrin protein; Metalloproteinase-like, disintegrin-like, and cysteine-rich protein 15; Metargidin; tMDC VI
Gene Aliases: ADAM15; MDC15; metargidin; tMDCVI
UniProt ID: (Human) Q13444, (Rat) Q9QYV0, (Mouse) O88839
Entrez Gene ID: (Human) 8751, (Rat) 57025, (Mouse) 11490
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support