Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: RLDPFFKQQA VCGNAKLEAG EECDCGTEQD CALIGETCCD IATCRFKAGS NCAEGPCCEN CLFMSKERMC RPSFEECDLP EYCNGSSASC PENHYVQTGH
ADAM2 is a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. ADAM2 is a subunit of an integral sperm membrane glycoprotein called fertilin, which plays an important role in sperm-egg interactions. This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This member is a subunit of an integral sperm membrane glycoprotein called fertilin, which plays an important role in sperm-egg interactions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a disintegrin and metalloproteinase; ADAM; ADAM 2; ADAMs; Cancer/testis antigen 15; CT15; Disintegrin and metalloproteinase domain-containing protein 2; Fertilin subunit beta; metalloendopeptidases; PH-30; PH30; PH30-beta
Gene Aliases: ADAM2; CRYN1; CRYN2; CT15; FTNB; PH-30b; PH30; PH30-beta
UniProt ID: (Human) Q99965
Entrez Gene ID: (Human) 2515
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support