Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: YKHPKYLELI LLFDQSRYRF VNNNLSQVIH DAILLTGIMD TYFQDVRMRI HLKALEVWTD FNKIRVGYPE LAEVLGRFVI YKKSVLNARL SSDWAHLYLQ
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This gene is testis-specific and contains a polymorphic region, resulting in isoforms with varying numbers of C-terminal repeats.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 30; ADAM; ADAM 30; ADAMs; Disintegrin and metalloproteinase domain-containing protein 30; metalloendopeptidases
Gene Aliases: ADAM30; svph4; UNQ2509/PRO5997
UniProt ID: (Human) Q9UKF2
Entrez Gene ID: (Human) 11085
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support