Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MFRLWLLLAG LCGLLASRPG FQNSLLQIVI PEKIQTNTND SSEIEYEQIS YIIPIDEKLY TVHLKQRYFL ADNFMIYLYN QGSMNTYSSD IQTQCYYQGN IEGYPDSMVT LSTCSGLRGI LQFENVSYGI EPLESAVEFQ HVLYKLKNED NDIAIFIDRS LKEQPMDDNI FISEKSEPAV PDLFPLYLEM HIVVDKTLYD YWGSDSMIVT NKVIEIVGLA NSMFTQFKVT IVLSSLELWS DENKISTVGE ADELLQKFLE WKQSYLNLRP HDIAYLLIYM DYPRYLGAVF PGTMCITRYS AGVALYPKEI TLEAFAVIVT QMLALSLGIS YDDPKKCQCS ESTCIMNPEV VQSNGVKTFS SCSLRSFQNF ISNVGVKCLQ NKPQMQKKSP KPVCGNGRLE GNEICDCGTE AQCGPASCCD FRTCVLKDGA KCYKGLCCKD CQILQSGVEC RPKAHPECDI AENCNGSSPE CGPDITLING LSCKNNKFIC YDGDCHDLDA RCESVFGKGS RNAPFACYEE IQSQSDRFGN CGRDRNNKYV FCGWRNLICG RLVCTYPTRK PFHQENGDVI YAFVRDSVCI TVDYKLPRTV PDPLAVKNGS QCDIGRVCVN RECVESRIIK ASAHVCSQQC SGHGVCDSRN KCHCSPGYKP PNCQIRSKGF SIFPEEDMGS IMERASGKTE NTWLLGFLIA LPILIVTTAI VLARKQLKKW FAKEEEFPSS ESKSEGSTQT YASQSSSEGS TQTYASQTRS ESSSQADTSK SKSEDSAEAY TSRSKSQDST QTQSSSN
This gene encodes a member of the disintegrin family of membrane-anchored proteins that play a role in diverse biological processes such as brain development, fertilization, tumor development and inflammation. This gene is predominantly expressed in the testis. The encoded protein undergoes proteolytic processing to generate a mature polypeptide comprised of an metalloprotease, disintegrin and epidermal growth factor-like domains. This gene is located in a cluster of other disintegrin and metallopeptidase family genes on chromosome 8. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq, Sep 2015]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a disintegrin and metalloprotease domain 32; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 32; ADAM; ADAM 32; ADAMs; Disintegrin and metalloproteinase domain-containing protein 32; FLJ26299; FLJ29004; metalloendopeptidases; metalloproteinase 12-like protein; testicular tissue protein Li 13
Gene Aliases: ADAM32; UNQ5982/PRO21340
UniProt ID: (Human) Q8TC27
Entrez Gene ID: (Human) 203102
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support