Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
In Western blots (under reducing and non-reducing conditions), approximately 20% cross-reactivity with the recombinant human (rh) ADAM9 Ectodomain is observed and no cross-reactivity with the Ectodomain of recombinant mouse ADAM10 and rhADAM8, 15, and 17 (TACE) is observed.
Reconstitute at 0.2 mg/mL in sterile PBS.
ADAM9 is a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This protein interacts with SH3 domain-containing proteins, binds mitotic arrest deficient 2 beta protein, and is also involved in TPA-induced ectodomain shedding of membrane-anchored heparin-binding EGF-like growth factor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a disintegrin and metalloprotease domain 9; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 9 (meltrin gamma); ADAM; ADAM 9; ADAM metallopeptidase domain 9 (meltrin gamma); adam9; ADAMs; Cellular disintegrin-related protein; cone rod dystrophy 9; Disintegrin and metalloproteinase domain-containing protein 9; MDC 9; Meltrin-gamma; metalloendopeptidases; Metalloprotease/disintegrin/cysteine-rich protein 9; Myeloma cell metalloproteinase; OTTHUMP00000216481; OTTHUMP00000216484; OTTHUMP00000216485
Gene Aliases: ADAM9; AU020942; CORD9; KIAA0021; MCMP; MDC9; mKIAA0021; MLTNG
UniProt ID: (Human) Q13443, (Mouse) Q61072
Entrez Gene ID: (Human) 8754, (Mouse) 11502
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support