Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: PDVPFNTKCP SGSCVMNQYL SSKFPKDFST SCRAHFERYL LSQKPKCLLQ APIPTNIMTT PVCGNHLLEV GEDCDCGSPK ECTNLCCEAL TCKLKPGTDC GGDAPNHTTE
ADAMDEC1 may play an important role in the control of the immune response and during pregnancy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a disintegrin and metalloproteinase; A disintegrin and metalloproteinase domain-like protein decysin-1; ADAM; ADAM DEC1; ADAM-like protein decysin-1; ADAM-like, decysin 1; ADAMs; decysin; disintegrin protease; metalloendopeptidases
Gene Aliases: ADAMDEC1; M12.219
UniProt ID: (Human) O15204
Entrez Gene ID: (Human) 27299
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support