Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: DGDWSPWGAW SMCSRTCGTG ARFRQRKCDN PPPGPGGTHC PGASVEHAVC ENLPCPKGLP SFRDQQCQAH DRLSPKKKGL LTAVVVDDKP CELYCSPLGK ESPLLVAD
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. ADAMTS family members share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The encoded preproprotein is proteolytically processed to generate the mature protein, which may promote breast cancer cell growth and survival. Mutations in this gene are associated with a Weill-Marchesani-like syndrome, which is characterized by lenticular myopia, ectopia lentis, glaucoma, spherophakia, and short stature. [provided by RefSeq, May 2016].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 17; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 17; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif, 17; ADAM-TS 17; ADAM-TS17; ADAMs; ADAMTS-17; ATS17; disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 17; FLJ16363; FLJ32769; metalloendopeptidases
Gene Aliases: ADAMTS17; AU023434
UniProt ID: (Human) Q8TE56
Entrez Gene ID: (Human) 170691, (Mouse) 233332
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support