Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 12
Antibody detects endogenous levels of total ADCY10.
SAC belongs to a distinct class of mammalian adenylyl cyclase that is soluble and insensitive to G protein or forskolin regulation. It is localized in the cytoplasm and is thought to function as a general bicarbonate sensor throughout the body. It may also play an important role in the generation of cAMP in spermatozoa, implying possible roles in sperm maturation through the epididymis, capacitation, hypermotility, and/or the acrosome reaction.The protein encoded by this gene belongs to a distinct class of mammalian adenylyl cyclase that is soluble and insensitive to G protein or forskolin regulation. It is localized in the cytoplasm and is thought to function as a general bicarbonate sensor throughout the body. It may also play an important role in the generation of cAMP in spermatozoa, implying possible roles in sperm maturation through the epididymis, capacitation, hypermotility, and/or the acrosome reaction. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3',5'-cyclic AMP synthetase; adenylate cyclase 10 (soluble); Adenylate cyclase homolog; Adenylate cyclase type 10; AH-related protein; ATP pyrophosphate-lyase; epididymis secretory sperm binding protein Li 7a; Germ cell soluble adenylyl cyclase; hsAC; RP1-313L4.2; sAC; soluble adenylyl cyclase; Testicular soluble adenylyl cyclase; testicular soluble adenylyl cyclase (SAC)
Gene Aliases: 4930431D04Rik; 4931412F17; ADCY10; HCA2; HEL-S-7a; hsAC; SAC; SACI; Sacy
UniProt ID: (Human) F5GWS5, (Rat) Q9Z286, (Mouse) Q8C0T9
Entrez Gene ID: (Human) 55811, (Rat) 59320, (Mouse) 271639
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support