Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: VTLEHLNGAY KVEEGDGDIR DPYLKQHLVK TYFVINPKGE RRSPQHLFRP RHTLDGAKMR ASVRMTRYLE SWGAAKPFAH LHHRDSMTTE NGKISTTDVP MGQHNFQNRT LRTKSQKKRF EEELNERMIQ AIDGINAQKQ WLKSEDIQRI SLLFYNKVLE KEYRATALPA F; Positive Samples: SH-SY5Y, Rat eye; Cellular Location: Cell membrane, Cytoplasm, Membrane, Multi-pass membrane protein
ADCY2 is a member of the family of adenylate cyclases, which are membrane-associated enzymes that catalyze the formation of the secondary messenger cyclic adenosine monophosphate(cAMP). This enzyme is insensitive to Ca(2+)/calmodulin, and is stimulated by the G protein beta and gamma subunit complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3',5'-cyclic AMP synthetase; AC2; adenylate cyclase 2 (brain); adenylate cyclase II; Adenylate cyclase type 2; Adenylate cyclase type II; Adenylyl cyclase 2; ATP pyrophosphate-lyase 2; type II adenylate cyclase
Gene Aliases: AC2; ADCY2; HBAC2; KIAA1060; mKIAA1060
UniProt ID: (Human) Q08462, (Rat) P26769
Entrez Gene ID: (Human) 108, (Mouse) 210044, (Rat) 81636
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support