Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSEETVPEAA SPPPPQGQPY FDRFSEDDPE YMRLRNRAAD LRQDFNLMEQ KKRVTMILQS PSFREELEGL IQEQMKKGNN SSNIWALRQI ADFMASTSHA VFPTSSMNVS MMTPINDLHT ADSLNLAKGE RLMRCKISSV YRLLDLYGWA QLSDTYVTLR VSKEQDHFLI SPKGVSCSEV TASSLIKVNI LGEVVEKGSS CFPVDTTGFC LHSAIYAARP DVRCIIHLHT PATAAVSAMK WGLLPVSHNA LLVGDMAYYD FNGEMEQEAD RINLQKCLGP TCKILVLRNH GVVALGDTVE EAFYKIFHLQ AACEIQVSAL SSAGGVENLI LLEQEKHRPH EVGSVQWAGS TFGPMQKSRL GEHEFEALMR MLDNLGYRTG YTYRHPFVQE KTKHKSEVEI PATVTAFVFE EDGAPVPALR QHAQKQQKEK TRWLNTPNAY LRVNVADEVQ RSMGSPRPKT TWMKADEVEK SSSGMPIRIE NPNQFVPLYT DPQEVLEMRN KIREQNRQDV KSAGPQSQLL ASVIAEKSRS PSTESQLMSK GDEDTKDDSE ETVPNPFSQL TDQELEEYKK EVERKKLELD GEKETAPEEP GSPAKSAPAS PVQSPAKEAE TKSPLVSPSK SLEEGTKKTE TSKAATTEPE TTQPEGVVVN GREEEQTAEE ILSKGLSQMT TSADTDVDTS KDKTESVTSG PMSPEGSPSK SPSKKKKKFR TPSFLKKSKK KEKVES
Adducins are heteromeric proteins composed of different subunits referred to as adducin alpha, beta and gamma. The three subunits are encoded by distinct genes and belong to a family of membrane skeletal proteins involved in the assembly of spectrin-actin network in erythrocytes and at sites of cell-cell contact in epithelial tissues. While adducins alpha and gamma are ubiquitously expressed, the expression of adducin beta is restricted to brain and hematopoietic tissues. Adducin, originally purified from human erythrocytes, was found to be a heterodimer of adducins alpha and beta. Polymorphisms resulting in amino acid substitutions in these two subunits have been associated with the regulation of blood pressure in an animal model of hypertension. Heterodimers consisting of alpha and gamma subunits have also been described. Structurally, each subunit is comprised of two distinct domains. The amino-terminal region is protease resistant and globular in shape, while the carboxy-terminal region is protease sensitive. The latter contains multiple phosphorylation sites for protein kinase C, the binding site for calmodulin, and is required for association with spectrin and actin. Various adducin beta mRNAs, alternatively spliced at 3'end and/or internally spliced and encoding different isoforms, have been described. The functions of all the different isoforms are not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adducin 2 (beta); Beta-adducin; Erythrocyte adducin subunit beta
Gene Aliases: ADD2; ADDB
UniProt ID: (Human) P35612
Entrez Gene ID: (Human) 119
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support