Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Immunogen sequence: DRIEERLPIL NQPSTQIVAN AKGAVTGAKD AVTTTVTGAK DSVASTITGV MDKTKGAVTG SVEKTKSVVS GSINTVLGSR MMQLVSSGVE NALTKSELLV EQYLPLTEEE LEKEAKKVEG FDLVQKPSYY VRLGSLSTKL HSRAYQQALS RVKEAKQKSQ QTISQLHSTV HLIEFARKNV YSANQKIQDA QDKLYLSWVE WKRSIGYDDT DESHCAEHIE SRTLAIARNL TQQLQTTCHT LLSNIQGVPQ NIQDQAKHMG VMAGDIYSVF RNAASFKEVS DSLLTSSKGQ LQKMKESLDD VMDYLVNNTP LNWLVGPFYP QLTESQNAQD QGAEMDKSSQ ETQRSEHKTH
Milk lipid globules from humans, cows and rats contain a protein identified as adipocyte differentiation-related protein (ADFP). It is associated with the globule surface membrane material. This protein, previously believed to be specific to adipocytes, is a major constituent of the globule surface and is present in a detergent-insoluble complex that contains butyrophilin and xanthine oxidase. ADFP (Adipophilin) is found in a wide range of cultured cell lines, including fibroblasts, endothelial and epithelial cells. In tissues, however, expression of adipophilin is restricted to specific cell types, such as lactating mammary epithelial cells, adrenal cortex cells, Sertoli and Leydig cells of the male reproductive system, and steatosis or fatty change hepatocytes in alcoholic liver cirrhosis. ADFP may be a possible new marker for the identification of specialized differentiated cells containing lipid droplets and for diseases associated with fat-accumulating cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adipophilin; adipose differentiation related protein; Adipose differentiation-related protein; ADRP; ADRP antibody; MGC10598; Perilipin-2
Gene Aliases: AA407157; ADFP; ADPH; ADRP; PLIN2
UniProt ID: (Human) Q99541, (Mouse) P43883
Entrez Gene ID: (Human) 123, (Mouse) 11520
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support