Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSTTGQVIRC KAAILWKPGA PFSIEEVEVA PPKAKEVRIK VVATGLCGTE MKVLGSKHLD LLYPTILGHE GAGIVESIGE GVSTVKPGDK VITLFLPQCG ECTSCLNSEG NFCIQFKQSK TQLMSDGTSR FTCKGKSIYH FGNTSTFCEY TVIKEISVAK IDAVAPLEKV CLISCGFSTG FGAAINTAKV TPGSTCAVFG LGGVGLSVVM GCKAAGAARI IGVDVNKEKF KKAQELGATE CLNPQDLKKP IQEVLFDMTD AGIDFCFEAI GNLDVLAGRA DSTSLNWLLI IWQRS
This gene encodes class V alcohol dehydrogenase, which is a member of the alcohol dehydrogenase family. Members of this family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. This gene is expressed in the stomach as well as in the liver, and it contains a glucocorticoid response element upstream of its 5' UTR, which is a steroid hormone receptor binding site. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alcohol dehydrogenase 6; aldehyde reductase
Gene Aliases: ADH-5; ADH6
UniProt ID: (Human) P28332
Entrez Gene ID: (Human) 130
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support