Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
In direct ELISAs, approximately 30% cross-reactivity with recombinant human ADNP is observed.
Reconstitute at 0.2 mg/mL in sterile PBS.
Vasoactive intestinal peptide is a neuroprotective factor that has a stimulatory effect on the growth of some tumor cells and an inhibitory effect on others. This gene encodes a protein that is upregulated by vasoactive intestinal peptide and may be involved in its stimulatory effect on certain tumor cells. The encoded protein contains one homeobox and nine zinc finger domains, suggesting that it functions as a transcription factor. This gene is also upregulated in normal proliferative tissues. Finally, the encoded protein may increase the viability of certain cell types through modulation of p53 activity. Alternatively spliced transcript variants encoding the same protein have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activity-dependent neuroprotective protein; Activity-dependent neuroprotector homeobox protein; NAP peptide
Gene Aliases: AA589558; Adnp; mKIAA0784
UniProt ID: (Mouse) A2BDX0, (Rat) Q9JKL8
Entrez Gene ID: (Mouse) 11538, (Rat) 64622
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support