Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total ADPGK.
ADPGK.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-dependent glucokinase; ADP-GK; ATP-dependent glucokinase; EC 2.7.1.147; RbBP-35
Gene Aliases: 2610017G09Rik; ADP-GK; ADPGK; C77577; PSEC0260; RGD1306103
UniProt ID: (Human) Q9BRR6, (Mouse) Q8VDL4
Entrez Gene ID: (Human) 83440, (Mouse) 72141, (Rat) 315722
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support