Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEKYVAAMV LSAAGDALGY YNGKWEFLQD GEKIHRQLAQ LGGLDALDVG RWRVSDDTVM HLATAEALVE AGKAPKLTQL YYLLAKHYQD CMEDMDGRAP GGASVHNAMQ LKPGKPNGWR IPFNSHEGGC GAAMRAMCIG LRFPHHSQLD TLIQVSIESG RMTHHHPTGY LGALASALFT AYAVNSRPPL QWGKGLMELL PEAKKYIVQS GYFVEENLQH WSYFQTKWEN YLKLRGILDG ESAPTFPESF GVKERDQFYT SLSYSGWGGS SGHDAPMIAY DAVLAAGDSW KELAHRAFFH GGDSDSTAAI AGCWWGVMYG FKGVSPSNYE KLEYRNRLEE TARALYSLGS KEDTVISL (1-357 aa encoded by BC063883)
Catalyzes the reverse reaction of mono-ADP-ribosylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [Protein ADP-ribosylarginine] hydrolase; ADP-ribose-L-arginine cleaving enzyme; ADP-ribosylarginine hydrolase; ADP-ribosylhydrolase ARH1
Gene Aliases: ADPRH; ADPRHA; ARH1
UniProt ID: (Human) P54922, (Mouse) P54923, (Rat) Q02589
Entrez Gene ID: (Human) 141, (Mouse) 11544, (Rat) 25371
Molecular Function:
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support