Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody specifically recognizes Arogenate dehydratase in Oryza sativa subspecies japonica (Rice).
Converts the prephenate produced from the shikimate-chorismate pathway into phenylalanine. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arogenate dehydratase/prephenate dehydratase 2, chloroplastic; AtADT2
Gene Aliases: ADT2; Arabidopsis thaliana arogenate dehydratase 2; arogenate dehydratase 2; AT3G07630; AtADT2; MLP3.8; PDT2
UniProt ID: (Arabidopsis) Q9SSE7
Entrez Gene ID: (Arabidopsis) 819954
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support