Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: VTDEQGIPIA NATISVSGIN HGVKTASGGD YWRILNPGEY RVTAHAEGYT PSAKTCNVDY DIGATQCNFI LARSNWKRIR EIMAMNGNRP IPHIDPSRPM TP
The adipocyte enhancer binding protein 1 (AEBP1) is a transcriptional repressor with carboxypeptidase (CP) activity. This protein binds to a regulatory sequence, adipocyte enhancer 1 (AE-1), located in the proximal promoter region of the adipose P2 (aP2) gene, which encodes the adipocyte fatty-acid binding protein. It is characterized as a member of the regulatory B-like CP family. This protein seems to be activated by a novel mechanism, whereby the direct binding of DNA enhances its protease activity. AEBP1 may play a role in differentiated vascular smooth muscle cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adipocyte enhancer-binding protein 1; AE-binding protein 1; Aortic carboxypeptidase-like protein; FLJ33612
Gene Aliases: ACLP; AEBP1
UniProt ID: (Human) Q8IUX7
Entrez Gene ID: (Human) 165
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support