Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DLKTHIVSI PLRGCEVIPG LDCKHPLTFR LLRNGQEVAV LEASSSEDMG RWIGILLAET GSSTDPEALH YDYIDVEMSA SVIQTAKQTF CFMNRRVISA NPYLGGTSNG YAHPSGTALH YDDVPCINGS LKGKKPPVAS NGVTGKGKTL SSQPKKADPA AVVKRTGSNA AQYKYGKNRV EADAKRLQTK EEELLKRKEA LRNRLAQLRK ERKDLRAAIE VNAGRKPQAI LEEKLKQLEE ECRQKEAERV SLELELTEVK ESLKKALAGG VTLGLAIEPK SGTSSPQSPV FRHRTLENSP ISSCDTSDTE GPVPVNSAAV LKKSQAAPGS SPCRGHVLRK AKEWELKNGT (382-730 aa encoded by BC032777)
The protein encoded by this gene is a Src binding partner. It may represent a potential modulator of actin filament integrity in response to cellular signals, and may function as an adaptor protein by linking Src family members and/or other signaling proteins to actin filaments. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 110 kDa actin filament-associated protein; Actin filament-associated protein 1; actin filament-associated protein, 110 kDa; AFAP-110; FLJ56849
Gene Aliases: 2600003E23Rik; 9630044L16Rik; AFAP; AFAP-110; AFAP1; AFAP110; AI848729; Kiaa3018; mKIAA3018
UniProt ID: (Human) Q8N556, (Mouse) Q80YS6, (Rat) Q8VH46
Entrez Gene ID: (Human) 60312, (Mouse) 70292, (Rat) 140935
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support