Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNREDRNVL RMKERERRNQ EIQQGEDAFP PSSPLFAEPY KVTSKEDKLS SRIQSMLGNY DEMKDFIGDR SIPKLVAIPK PTVPPSADEK SNPNFFEQRH GGSHQSSKWT PVGPAPSTSQ SQKRSSGLQS GHSSQRTSAG SSSGTNSSGQ RHDRESYNNS GSSSRKKGQH GSEHSKSRSS SPGKPQAVSS LNSSHSRSHG NDHHSKEHQR SKSPRDPDAN WDSPSRVPFS SGQHSTQSFP PSLMSKSNSM LQKPTAYVRP MDGQESMEPK LSSEHYSSQS HGNSMTELKP SSKAHLTKLK IPSQPLDASA SGDVSCVDEI LKEMTHSWPP PLTAIHTPC (1-338 aa encoded by BC063007 )
The protein encoded by this gene belongs to the AF4 family of transcription factors involved in leukemia. It is a component of the positive transcription elongation factor b (P-TEFb) complex. A chromosomal translocation involving this gene and MLL gene on chromosome 11 is found in infant acute lymphoblastic leukemia with ins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AF4/FMR2 family member 4; AF4/FMR2 family, member 4; ALL1 fused gene from 5q31; ALL1-fused gene from chromosome 5q31 protein; Major CDK9 elongation factor-associated protein; Protein AF-5q31
Gene Aliases: AF5Q31; AFF4; Alf4; CHOPS; HSPC092; Laf4l; MCEF
UniProt ID: (Human) Q6P592, (Mouse) Q9ESC8
Entrez Gene ID: (Human) 27125, (Mouse) 93736
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support