Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKLLKLTGFI FFLFFLTESL TLPTQPRDIE NFNSTQKFIE DNIEYITIIA FAQYVQEATF EEMEKLVKDM VEYKDRCMAD KTLPECSKLP NNVLQEKICA MEGLPQKHNF SHCCSKVDAQ RRLCFFYNKK SDVGFLPPFP TLDPEEKCQA YESNRESLLN HFLYEVARRN PFVFAPTLLT VAVHFEEVAK SCCEEQNKVN CLQTRAIPVT QYLKAFSSYQ KHVCGALLKF GTKVVHFIYI AILSQKFPKI EFKELISLVE DVSSNYDGCC EGDVVQCIRD TSKVMNHICS KQDSISSKIK ECCEKKIPER GQCIINSNKD DRPKDLSLRE GKFTDSENVC QERDADPDTF FAKFTFEYSR RHPDLSIPEL LRIVQIYKDL LRNCCNTENP PGCYRYAEDK FNETTEKSLK MVQQECKHFQ NLGKDGLKYH YLIRLTKIAP QLSTEELVSL GEKMVTAFTT CCTLSEEFAC VDNLADLVFG ELCGVNENRT INPAVDHCCK TNFAFRRPCF ESLKADKTYV PPPFSQDLFT FHADMCQSQN EELQRKTDRF LVNLVKLKHE LTDEELQSLF TNFANVVDKC CKAESPEVCF NEESPKIGN
This gene is a member of the albumin gene family, which is comprised of four genes that localize to chromosome 4 in a tandem arrangement. These four genes encode structurally-related serum transport proteins that are known to be evolutionarily related. The protein encoded by this gene is regulated developmentally, expressed in the liver and secreted into the bloodstream.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Afamin; Alpha-Alb; Alpha-albumin; MGC125338; MGC125339
Gene Aliases: AFM; ALB2; ALBA; ALF
UniProt ID: (Human) P43652
Entrez Gene ID: (Human) 173
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support