Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LDVAHVHEHC CRGDVLDCLQ DGEKIMSYIC SQQDTLSNKI TECCKLTTLE RGQCIIHAEN DEKPEGLSPN LNRFLGDRDF NQFSSGEKNI FLASFVHEYS RRHPQLAVSV ILRVAKGYQE LLEKCFQTEN PLECQDKGEE ELQKYIQESQ ALAKRSCGLF QKLGEYYLQN AFLVAYTKKA PQLTSSELMA ITRKMAATAA TCCQLSEDKL LACGEGAADI IIGHLCIRHE MTPVNPGVGQ CCTSSYANRR PCFSSLVVDE TYVPPAFSDD KFIFHKDLCQ AQGVALQTMK QEFLINLVKQ KPQITEEQLE AVIADFSGLL EKCCQGQEQE VCFAEEGQKL ISKTRAALGV
AFP (Alpha Fetoprotein) is a major plasma protein produced by the yolk sac, intestinal tract and the liver during fetal life. AFP expression in adults is often associated with hepatoma or teratoma. However, hereditary persistence of AFP may also be found in individuals with no obvious pathology. The protein is thought to be the fetal counterpart of serum albumin, and AFP and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. AFP is found in monomeric, dimeric, and trimeric forms. AFP can also bind to copper ions, nickel ions, fatty acids and bilirubin. The level of AFP in amniotic fluid is used to measure renal loss of protein to screen for spina bifida and anencephaly. High level of serum AFP has been identified in patients with hepatocellular carcinomas (HCC), teratoblastoma, colorectal cancer, pancreatic cancer, and germ cell neoplasms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a fetoglobulin; a fetoprotein; Alpha fetoglobulin; Alpha-1-fetoprotein; Alpha-fetoglobulin; Alpha-fetoprotein; fetoglobulin; fetoprotein; α fetoglobulin; α fetoprotein
Gene Aliases: AFP; AFPD; FETA; HPAFP
UniProt ID: (Human) P02771
Entrez Gene ID: (Human) 174
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support