Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MLRLLASGCA RGPGPGVGAR PAAGLFHPGR RQSRQASDAP RNQPPSPEFV ARPVGVCSMM RLPVQTSPEG LDAAFIGVPL DTGTSNRPGA RFGPRRIREE SVMLRTVNPS TGALPFQSLM VADLGDVNVN LYNLQDSCRR IQEAYEKIVA AGCIPLTLGG DHTITYPILQ AMAKKHGPVG LLHVDAHTDT TDKALGEKLY HGAPFRRCVD EGLLDCKRVV QIGIRGSSTT LDPYRYNRSQ GFRVVLAEDC WMKSLVPLMG EVRQQMGGKP IYISFDIDAL DPAYAPGTGT PEIAGLTPSQ ALEIIRGCQG LNVMGCDLVE VSPPYDLSGN TALLAANLLF EMLCALPKVT TV
AGMAT (Agmatinase) hydrolyzes agmatine to putrescine and urea. It is highly expressed in liver and kidney and is also found in skeletal muscle, fetal liver, brain, testis, skin and the gastrointestinal tract.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: agmatine ureohydrolase (agmatinase); Arginase, mitochondrial; AUH; Guanidino acid hydrolase, mitochondrial; Guanidinobutyrase, mitochondrial; Guanidinopropionase, mitochondrial
Gene Aliases: AGMAT; GDAH
UniProt ID: (Human) Q9BSE5
Entrez Gene ID: (Human) 79814
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support