Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total PLCH.
This gene encodes a member of the lysophosphatidic acid acyltransferase protein family. The encoded protein is an enzyme which catalyzes the conversion of glycerol-3-phosphate to lysophosphatidic acid in the synthesis of triacylglycerol. Multiple alternatively spliced variants, encoding the same protein, have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-acyl-sn-glycerol-3-phosphate O-acyltransferase 10; 1-acyl-sn-glycerol-3-phosphate O-acyltransferase 9; 1-acylglycerol-3-phosphate O-acyltransferase 8; 1-acylglycerol-3-phosphate O-acyltransferase 9; 1-AGP acyltransferase 9; 1-AGPAT 9; Acyl-CoA:glycerol-3-phosphate acyltransferase 3; AGPAT 10; endoplasmic reticulum associated GPAT; Glycerol-3-phosphate acyltransferase 3; GPAT-3; GPAT3; hGPAT3; LPAAT-theta; Lung cancer metastasis-associated protein 1; Lysophosphatidic acid acyltransferase theta; lysophosphatidic acid acyltransferase, theta; MAG-1; mGPAT3; testis secretory sperm-binding protein Li 213e
Gene Aliases: 1-AGPAT; 1-AGPAT 9; 4933407I02Rik; 4933408F15; 9; A230097K15Rik; AGPAT 10; AGPAT10; AGPAT8; AGPAT9; GPAT-3; GPAT3; HMFN0839; LPAAT-theta; MAG1; mGPAT3; UNQ2753/PRO6492
UniProt ID: (Rat) Q4V8J4, (Mouse) Q8C0N2, (Human) Q53EU6
Entrez Gene ID: (Rat) 305166, (Mouse) 231510, (Human) 84803
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support