Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MELPAVNLK VILLGHWLLT TWGCIVFSGS YAWANFTILA LGVWAVAQRD SIDAISMFLG GLLATIFLDI VHISIFYPRV SLTDTGRFGV GMAILSLLLK PLSCCFVYHM YRERGGFLGS SQDRSAYQTI DSAEAPADPF AVPEGRSQDA RGY (1-152 aa encoded by BC017328)
This gene encodes a transmembrane protein localized to the plasma membrane and perinuclear vesicular structures. The gene product interacts with the angiotensin II type I receptor and negatively regulates angiotensin II signaling. Alternative splicing of this gene generates multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: angiotensin II receptor-associated protein; angiotensin II, type I receptor-associated protein; AT1 receptor-associated protein; ATI receptor-associated protein; Type-1 angiotensin II receptor-associated protein
Gene Aliases: AGTRAP; ATRAP
UniProt ID: (Human) Q6RW13
Entrez Gene ID: (Human) 57085
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support