Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Samples: 22Rv1, SW480, HepG2, U-937, SW620, MCF7, K-562, Mouse liver; Cellular Location: Cytoplasm, Melanosome
Immunogen sequence: AMEGYEVTTM DEACQEGNIF VTTTGCIDII LGRHFEQMKD DAIVCNIGHF DVEIDVKWLN ENAVEKVNIK PQVDRYRLKN GRRIILLAEG RLVNLGCAMG HPSFVMSNSF TNQVMAQIEL WTHPDKYPVG VHFLPKKLDE AVAEAHLGKL NVKLTKLTEK QAQYLGMSCD GPFKPDHYRY
S-adenosylhomocysteine hydrolase (AHCY) catalyzes the reversible hydrolysis of S-adenosylhomocysteine (AdoHcy) to adenosine (Ado) and L-homocysteine (Hcy). Thus, it regulates the intracellular S-adenosylhomocysteine (SAH) concentration thought to be important for transmethylation reactions. Deficiency in this protein is one of the different causes of hypermethioninemia. S-adenosylhomocysteine hydrolase belongs to the adenosylhomocysteinase family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acetylation; Adenosylhomocysteinase; AdoHcyase; CUBP; Hydrolase; Liver copper-binding protein; NAD; Polymorphism; S-adenosyl-L-homocysteine hydrolase; S-adenosylhomocysteine hydrolase
Gene Aliases: AA987153; adoHcyase; AHCY; AL024110; CuBP; SAHH
UniProt ID: (Human) P23526, (Mouse) P50247, (Rat) P10760
Entrez Gene ID: (Human) 191, (Mouse) 269378, (Rat) 29443
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support