Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: APHGPGLIYR QPNCDDPETE EAALVAIDYI NQNLPWGYKH TLNQIDEVKV WPQQPSGELF EIEIDTLETT CHVLDPTPVA RCSVRQLKEH AVEGDCDFQL LKLDGKFSVV YAKCDSSPDS AEDVRKVCQD CPLLAPLNDT RVVHAAKAAL AAFNAQNNGS NFQLEEISRA QLVPLPPSTY VEFTVSGTDC VAKEATEAAK CNLLAEKQYG FCKATLSEKL GGAEVAVTCT VFQTQPVTSQ PQPEGANEAV PTPVVDPDAP PSPPLGAPGL PPAGSPPDSH VLLAAPPGHQ LHRAHYDLRH TFMGVVSLGS PSGEVSHPRK TRTVVQPSVG AAAGPVVPPC PGRIRHFKV
Alpha2-HS glycoprotein (AHSG), a glycoprotein present in the serum, is synthesized by hepatocytes. The AHSG molecule consists of two polypeptide chains, which are both cleaved from a proprotein encoded from a single mRNA. It is involved in several functions, such as endocytosis, brain development and the formation of bone tissue. The protein is commonly present in the cortical plate of the immature cerebral cortex and bone marrow hemopoietic matrix, and it has therefore been postulated that it participates in the development of the tissues. However, its exact significance is still obscure.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-2-HS-glycoprotein; Alpha-2-Z-globulin; Ba-alpha-2-glycoprotein; fetuin A; Fetuin-A; FetuinA
Gene Aliases: A2HS; AHS; AHSG; FETUA; HSGA; PRO2743
UniProt ID: (Human) P02765
Entrez Gene ID: (Human) 197
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support