Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AIM2 is a member of the IFI20X /IFI16 family. It plays a putative role in tumorigenic reversion and may control cell proliferation. Interferon-gamma induces expression of AIM2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Absent in melanoma 2; Ifi-210; Interferon-inducible protein 210; Interferon-inducible protein AIM2; Interferon-inducible protein p210; RP11-520H16.3
Gene Aliases: AIM2; Gm1313; Ifi210; PYHIN4
UniProt ID: (Human) O14862, (Mouse) Q91VJ1
Entrez Gene ID: (Human) 9447, (Mouse) 383619
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support