Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MADIIARLR EDGIQKRVIQ EGRGELPDFQ DGTKATFHYR TLHSDDEGTV LDDSRARGKP MELIIGKKFK LPVWETIVCT MREGEIAQFL CDIKHVVLYP LVAKSLRNIA VGKDPLEGQR HCCGVAQMRE HSSLGHADLD ALQQNPQPLI FHMEMLKVES PGTYQQDPWA MTDEEKAKAV PLIHQEGNRL YREGHVKEAA AKYYDAIACL KNLQMKEQPG SPEWIQLDKQ ITPLLLNYCQ CKLVVEEYYE VLDHCSSILN KYDDNVKAYF KRGKAHAAVW NAQEAQADFA KVLELDPALA PVVSRELRAL EARIRQKDEE DKARFRGIFS H (1-330 aa encoded by BC104797)
The protein encoded by this gene is a receptor for aryl hydrocarbons and a ligand-activated transcription factor. The encoded protein is found in the cytoplasm as part of a multiprotein complex, but upon binding of ligand is transported to the nucleus. This protein can regulate the expression of many xenobiotic metabolizing enzymes. Also, the encoded protein can bind specifically to and inhibit the activity of hepatitis B virus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AH receptor-interacting protein; AIP; Aryl-hydrocarbon receptor-interacting protein; HBV X-associated protein 2; HBV-X associated protein; Immunophilin homolog ARA9; Immunophilin XAP2; XAP-2
Gene Aliases: AA408703; AIP; ARA9; AW476050; D19Bwg1412e; FKBP16; FKBP37; SMTPHN; XAP-2; XAP2
UniProt ID: (Human) O00170, (Mouse) O08915, (Rat) Q5FWY5
Entrez Gene ID: (Human) 9049, (Mouse) 11632, (Rat) 282827
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support