Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a protein containing 9 WD repeats. WD repeats are approximately 30- to 40-amino acid domains containing several conserved residues, mostly including a trp-asp at the C-terminal end. WD domains are involved in protein-protein interactions. The encoded protein may help induce the disassembly of actin filaments. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Actin-interacting protein 1; AIP1; epididymis secretory protein Li 52; NORI-1; WD repeat protein 1; WD repeat-containing protein 1; WD40 repeat protein 1
Gene Aliases: AIP1; D5Wsu185e; HEL-S-52; NORI-1; rede; WDR1
UniProt ID: (Human) A8MPU4, (Rat) Q5RKI0, (Mouse) O88342
Entrez Gene ID: (Human) 9948, (Rat) 360950, (Mouse) 22388
Molecular Function:
non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support