This antibody can recognize all the isoforms of AKAP7.
Immunogen sequence: MGQLCCFPF SRDEGKISEK NGGEPDDAEL VRLSKRLVEN AVLKAVQQYL EETQNKNKPG EGSSVKTEAA DQNGNDNENN RK (1-81 aa encoded by BC016927)
This gene encodes a member of the A-kinase anchoring protein family, a group of functionally related proteins that bind to a regulatory subunit of cAMP-dependent protein kinase A and target the enzyme to specific subcellular compartments. AKAPs have a common RII-binding domain, but contain different targeting motifs responsible for directing PKA to distinct intracellular locations. Three alternatively spliced transcript variants encoding different isoforms have been described. Additional variants exist, but their full-length natures have not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A kinase (PRKA) anchor protein 7; A-kinase anchor protein 18; A-kinase anchor protein 18 kDa; A-kinase anchor protein 18 {ECO:0000250|UniProtKB:F8VQ58}; A-kinase anchor protein 7; A-kinase anchor protein 7 isoform alpha; A-kinase anchor protein 7 isoform gamma; A-kinase anchor protein 7 isoforms alpha and beta; A-kinase anchor protein 7 isoforms delta and gamma; A-kinase anchor protein 9 kDa; A-kinase anchoring protein 18 ,isoform delta; AKAP 18; AKAP 9; AKAP-18; AKAP-18 {ECO:0000250|UniProtKB:F8VQ58}; AKAP-7 isoform alpha; AKAP-7 isoform gamma; AKAP-7 isoforms alpha and beta; AKAP-7 isoforms delta and gamma; akap7; akap7 {ECO:0000312|RGD:1303071}; PRKA7 isoform alpha; PRKA7 isoform gamma; PRKA7 isoforms alpha/beta; PRKA7 isoforms delta and gamma; protein kinase A anchoring protein 7; Protein kinase A-anchoring protein 7 isoform alpha; Protein kinase A-anchoring protein 7 isoform gamma; Protein kinase A-anchoring protein 7 isoforms alpha/beta; Protein kinase A-anchoring protein 7 isoforms delta and gamma
Gene Aliases: 6430401D08; AI662165; AKAP-18; AKAP15; AKAP18; AKAP18d; AKAP7; BB170514
UniProt ID: (Human) O43687, (Mouse) O55074, (Rat) Q6JP77
Entrez Gene ID: (Human) 9465, (Mouse) 432442, (Rat) 361458
Molecular Function:
RNA metabolism protein
RNA processing factor
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support