Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKERKRHLG DTKHFCPVVL KENFILQPGN TEEAAKYREK IYYFSSAEAK EKFLEHPEDY VAHEEPLKAP PLRICLVGPQ GSGKTMCGRQ LAEKLNIFHI QFEEVLQEKL LLKTEKKVGP EFEEDSENEQ AAKQELEELA IQANVKVEEE NTKKQLPEVQ LTEEEEVIKS SLMENEPLPP EILEVILSEW WLKEPIRSTG FILDGFPRYP EEAQFLGDRG FFPDAAVFIQ VDDQDIFDRL LPAQIEKWKL KQKKKLERKK LIKDMKAKIR VDTIAKRRAE LILERD (921-1205 aa encoded by BC153055 )
Involved in maintaining the homeostasis of cellular nucleotides by catalyzing the interconversion of nucleoside phosphates. Has both nucleoside monophosphate and diphosphate kinase activities. Catalyzes the phosphorylation of AMP, dAMP, CMP and dCMP with ATP as phosphate donor and of CMP with GTP as phosphate donor. Also catalyzes the production of ATP, CTP, GTP, UTP, dATP, dCTP, dGTP and TTP from the corresponding diphosphate substrates with either ATP or GTP as phosphate donor. Shows substrate preference of CDP > UDP > ADP > GDP > TDP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenylate kinase 9; adenylate kinase domain containing 1; adenylate kinase domain containing 2; Adenylate kinase domain-containing protein 1; Adenylate kinase domain-containing protein 2; RP1-70A9.1
Gene Aliases: AK 9; AK9; AKD1; AKD2; C6orf199; C6orf224; dJ70A9.1
UniProt ID: (Human) Q5TCS8
Entrez Gene ID: (Human) 221264
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support