Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TNQVECHPYL TQEKLIQYCH SKGISVTAYS PLGSPDRPSA KPEDPSLLED
For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aldehyde reductase; Aldose reductase-related protein 2; AR; fibroblast growth factor regulated protein; Fibroblast growth factor-regulated protein; Protein FR-1
Gene Aliases: Akr1b8; Fgfrp; Fgrp; FR-1
UniProt ID: (Mouse) P45377
Entrez Gene ID: (Mouse) 14187
Molecular Function:
oxidoreductase
reductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support