Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
ALAS-1 is a nuclear-encoded mitochondrial matrix enzyme which catalyzes the condensation of glycine with succinyl-CoA to form delta-amino levulinate, CO2 and CoA. It regulates the first and rate-limiting step of heme biosynthetic pathway. It is one of the two isoforms of ALAS and is a pyridoxal-5' phosphate dependant housekeeping enzyme. It is ubiquitously expressed and is responsible of providing heme for cytochromes and other hemoproteins. ALAS1 in liver undergoes negative feedback regulation by heme. Transcription of ALAS1 gene is stimulated by cAMP and respiratory uncoupling whereas Phorbol esters and insulin repress ALAS1 gene expression. ALAS1 is suggested to be involved in reciprocal regulation of Heme biosynthesis and circadian clock suggesting a potential target for treatment of circadian disorders. ALAS1 gene expression is down regulated in Acute Liver failure resulting in altered heme metabolism and liver function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5-aminolevulinate synthase, non-specific, mitochondrial; 5-aminolevulinic acid synthase 1; ALAS-H; aminolevulinate, delta-, synthase 1; Delta-ALA synthase 1; Delta-aminolevulinate synthase 1; migration-inducing protein 4
Gene Aliases: ALAS; ALAS-H; ALAS1; ALAS3; ALASH; MIG4; OK/SW-cl.121
UniProt ID: (Human) P13196
Entrez Gene ID: (Human) 211
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support