Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DDTVSTLYN RFLFPEGIKG MVQAVRLIAE GKAPRLPQPE EGATYEGIQK KETAKINWDQ PAEAIHNWIR GNDKVPGAWT EACEQKLTFF NSTLNTSGLV PEGDALPIPG AHRPGVVTKA GLILFGNDDK MLLVKNIQLE DGKMILASNF FKGAASSVLE LTEAELVTAE AVRSVWQRIL PKVLEVEDST DFFKSGAASV DVVRLVEEVK ELCDGLELEN EDVYMASTFG DFIQLLVRKL RGDDEEGECS IDYVEMAVNK RTVRMPHQLF IGGEFVDAEG AKTSETINPT DGSVICQVSL AQVTDVDKAV AAAKDAFENG RWGKISARDR GRLMYRAPPS PSTRPDPTAT (157-505 aa encoded by BC027241)
ALDH1L1 catalyzes the conversion of 10-formyltetrahydrofolate, NADP, and water to tetrahydrofolate, NADPH, and carbon dioxide. The encoded protein belongs to the aldehyde dehydrogenase family and is responsible for formate oxidation in vivo. Deficiencies in this gene can result in an accumulation of formate and subsequent methanol poisoning.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 10-formyltetrahydrofolate dehydrogenase; 10-FTHFDH; 10fTHF; 10FTHFDH; aldehyde dehydrogenase 1 family, member L1; Aldehyde dehydrogenase family 1 member L1; Cytosolic 10-formyltetrahydrofolate dehydrogenase; DKFZp781N0997; FBP-CI; formyltetrahydrofolate dehydrogenase; MER57B-ALDH1L1
Gene Aliases: 10-fTHF; 10-FTHFDH; 1810048F20Rik; ALDH1L1; FDH; FTHFD; Neut2
UniProt ID: (Human) O75891, (Rat) P28037, (Mouse) Q8R0Y6
Entrez Gene ID: (Human) 10840, (Rat) 64392, (Mouse) 107747
Molecular Function:
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
RNA processing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support