Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLLPAPALRR ALLSRPWTGA GLRWKHTSSL KVANEPVLAF TQGSPERDAL QKALKDLKGR MEAIPCVVGD EEVWTSDVQY QVSPFNHGHK VAKFCYADKS LLNKAIEAAL AARKEWDLKP IADRAQIFLK AADMLSGPRR AEILAKTMVG QGKTVIQAEI DAAAELIDFF RFNAKYAVEL EGQQPISVPP STNSTVYRGL EGFVAAISPF NFTAIGGNLA GAPALMGNVV LWKPSDTAML ASYAVYRILR EAGLPPNIIQ FVPADGPLFG DTVTSSEHLC GINFTGSVPT FKHLWKQVAQ NLDRFHTFPR LAGECGGKNF HFVHRSADVE SVVSGTLRSA FEYGGQKCSA CSRLYVPHSL WPQIKGRLLE EHSRIKVGDP AEDFGTFFSA VIDAKSFARI KKWLEHARSS PSLTILAGGK CDDSVGYFVE PCIVESKDPQ EPIMKEEIFG PVLSVYVYPD DKYKETLQLV DSTTSYGLTG AVFSQDKDVV QEATKVLRNA AGNFYINDKS TGSIVGQQPF GGARASGTND KPGGPHYILR WTSPQVIKET HKPLGDWSYA YMQ
ALDH4A1 belongs to the aldehyde dehydrogenase family of proteins. This enzyme is a mitochondrial matrix NAD-dependent dehydrogenase which catalyzes the second step of the proline degradation pathway, converting pyrroline-5-carboxylate to glutamate. Deficiency of this enzyme is associated with type II hyperprolinemia, an autosomal recessive disorder characterized by accumulation of delta-1-pyrroline-5-carboxylate (P5C) and proline.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aldehyde dehydrogenase 4 family, member A1; Aldehyde dehydrogenase family 4 member A1; Delta-1-pyrroline-5-carboxylate dehydrogenase, mitochondrial; L-glutamate gamma-semialdehyde dehydrogenase; mitochondrial delta-1-pyrroline 5-carboxylate dehydrogenase; P5C dehydrogenase; RP11-128M10.1
Gene Aliases: ALDH4; ALDH4A1; P5CD; P5CDH
UniProt ID: (Human) P30038
Entrez Gene ID: (Human) 8659
Molecular Function:
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support